Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Thermo Scientific Chemicals
Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
CAS: 1321-74-0 | C10H10 | 130.19 g/mol
Have Questions?
Catalog NumberQuantity
ALF042804.22100 g
Catalog number ALF042804.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDivinylbenzene
Name NoteMixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol
CAS1321-74-0
Health Hazard 1H227-H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
View more
Divinylbenzene is used as a reactive monomer in polyester resins and for the synthesis of highly crosslinked macroporous styrene as well as a crosslinking agent. It acts as an intermediate in the production of ion exchange resin. Further, it is used in making rubber, petroleum production and polymeric core shell nanoparticles. In addition, it is used to synthesize monodispersed poly divinylbenzene microspheres.