Copper gauze, 20 mesh woven from 0.41mm (0.016in) dia wire
Copper gauze, 20 mesh woven from 0.41mm (0.016in) dia wire
Copper gauze, 20 mesh woven from 0.41mm (0.016in) dia wire
Copper gauze, 20 mesh woven from 0.41mm (0.016in) dia wire
Thermo Scientific Chemicals
Copper gauze, 20 mesh woven from 0.41mm (0.016in) dia wire
Wire Cloth | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF042777.HZ300 x 1200 mm
Catalog number ALF042777.HZ
Price (MYR)
832.00
EA
Add to cart
Quantity:
300 x 1200 mm
Request bulk or custom format
Price (MYR)
832.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
AppearanceCopper colored guaze
Recommended StorageAmbient temperatures
Quantity300 x 1200 mm
View more
Filtration, electrodes