Silica gel, TLC high purity grade, without binder
Silica gel, TLC high purity grade, without binder
Silica gel, TLC high purity grade, without binder
Silica gel, TLC high purity grade, without binder
Thermo Scientific Chemicals
Silica gel, TLC high purity grade, without binder
For TLC Plate Preparation | CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF042756.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSilica gel
CAS63231-67-4
ColorWhite
Health Hazard 1H373: May cause damage to organs through prolonged or repeated exposure.
Recommended StorageAmbient temperatures
View more
Silica gel, TLC high purity grade, without binder is employed for thin layer chromatography plate preparation.