Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g
Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g
Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g
Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g
Thermo Scientific Chemicals
Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g
For Adhesives, Sealants, Silicones | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF042737.A11 kg
Catalog number ALF042737.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilicon(IV) oxide
Melting Point1710°C
CAS7631-86-9
ColorWhite
View more
Silicon(IV) oxide, amorphous is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used to manufacture adhesives, sealants and silicones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
S.A. 85 to 115m2/g
RUO – Research Use Only