Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%
Thermo Scientific Chemicals
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%
CAS: 135236-72-5 | C10H18CaO6 | 274.33 g/mol
Have Questions?
Catalog NumberQuantity
ALF042721.1850 g
Catalog number ALF042721.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium 3-hydroxy-3-methylbutyrate hydrate
CAS135236-72-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Calcium 3-hydroxy-3-methylbutyrate hydrate is a nutritional supplement, which represents a calcium salt of beta-hydroxy beta-methylbutyrate (HMB). The latter is a leucine metabolite produced in the body through oxidation of the ketoacid of leucine (ketoisocaproate, KIC). Because leucine plays an important regulatory role in protein metabolism and due to the conversion of leucine to KIC, several researchers have presumed that KIC is the active ingredient responsible for most of the beneficial effects of leucine on protein metabolism.