n-Octyltrimethoxysilane, 97+%
n-Octyltrimethoxysilane, 97+%
n-Octyltrimethoxysilane, 97+%
n-Octyltrimethoxysilane, 97+%
Thermo Scientific Chemicals
n-Octyltrimethoxysilane, 97+%
CAS: 3069-40-7 | C11H26O3Si | 234.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF042698.22
Price (MYR)
2,225.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,225.00
EA
Add to cart
Specifications
Boiling Point75°C (0.1mmHg)
Chemical Name or Materialn-Octyltrimethoxysilane
CAS3069-40-7
ColorColorless to Yellow
Health Hazard 1H315-H319-H331
View more
n-Octyltrimethoxysilane is used in medicine field, electronic industry and polymer material. It is also used as surface modifier.