Potassium chloride, 0.117M, Conductivity Standard
Potassium chloride, 0.117M, Conductivity Standard
Potassium chloride, 0.117M, Conductivity Standard
Potassium chloride, 0.117M, Conductivity Standard
Thermo Scientific Chemicals
Potassium chloride, 0.117M, Conductivity Standard
CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF042696.K21 L
Catalog number ALF042696.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium chloride
Name Note15, 000μmho at 25°C
CAS7447-40-7
CAS7732-18-5