Copper powder, spherical, APS 10 micron, 99.9% (metals basis)
Copper powder, spherical, APS 10 micron, 99.9% (metals basis)
Copper powder, spherical, APS 10 micron, 99.9% (metals basis)
Copper powder, spherical, APS 10 micron, 99.9% (metals basis)
Thermo Scientific Chemicals
Copper powder, spherical, APS 10 micron, 99.9% (metals basis)
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF042689.30
Price (MYR)
898.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
898.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
Health Hazard 1H228-H315-H319-H335-H500
AppearanceCopper colored powder
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c