Platinum Iridium wire, 1.0mm (0.04in) dia
Platinum Iridium wire, 1.0mm (0.04in) dia
Thermo Scientific Chemicals
Platinum Iridium wire, 1.0mm (0.04in) dia
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
5 cm
25 cm
Catalog number ALF042678.BY
Price (MYR)
51,856.00
EA
Add to cart
Quantity:
100 cm
Request bulk or custom format
Price (MYR)
51,856.00
EA
Add to cart
Specifications
Chemical Name or MaterialPlatinum Iridium wire
Name Note1.0mm (0.04 in.) dia.
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 90:10 wt%
Recommended StorageAmbient temperatures
Quantity100 cm