Polyethylene glycol 12,000
Polyethylene glycol 12,000
Polyethylene glycol 12,000
Polyethylene glycol 12,000
Thermo Scientific Chemicals
Polyethylene glycol 12,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF042635.30250 g
Catalog number ALF042635.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene Glycol
CAS25322-68-3
Physical FormPowder
Melting Point64°C to 65°C
Recommended StorageAmbient temperatures
View more
Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.