Polyethylene, low density, ∣<400 micron
Polyethylene, low density, &mid;&lt;400 micron
Polyethylene, low density, &mid;&lt;400 micron
Polyethylene, low density, &mid;&lt;400 micron
Thermo Scientific Chemicals
Polyethylene, low density, ∣<400 micron
CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF042607.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene
Name NoteLow density, ≤400 micron
CAS9002-88-4
Physical FormPowder
Recommended StorageAmbient temperatures
View more
Polyethylene is used in injection and blow molded toys, housewares, lids, injection molded car seats, Hot melt coating for paper, additive in cast moldings, candles, oil-based inks and hot melt adhesives. It is also used as an additive, lubricant in rubber processing, extrusion and calendaring aid for PVC and dispersing aid for color concentrates. Bearings, gears, bushings and other moving parts.