Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF042583.031 g
Catalog number ALF042583.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZirconium (IV) tert-butoxide
CAS2081-12-1
Health Hazard 1H226
Health Hazard 2GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c