Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000
Thermo Scientific Chemicals
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000
CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Have Questions?
Catalog NumberQuantity
ALF042490.K21 L
Catalog number ALF042490.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPolydimethylsiloxane, trimethylsiloxy terminated
ColorColorless
Name NoteM.W. 14,000
CAS9016-00-6
Physical FormViscous Liquid
View more
Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant, is used in coating auxiliary agents. It is also used as pharmaceutical intermediates.