Buffer solution, pH 9.18 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 9.18 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals
Buffer solution, pH 9.18 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Have Questions?
Catalog NumberQuantity
ALF042422.K21 L
Catalog number ALF042422.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialBuffer solution, pH 9.18 (±0.01 at 25°C)
Name NoteNIST Traceable
Health Hazard 1H290
Health Hazard 3P234-P390