Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals
Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF042421.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialBuffer solution, pH 9.00 (±0.01 at 25°C)
Name NoteNIST Traceable
Recommended StorageAmbient temperatures
TSCAYes
Physical FormLiquid
View more
Buffer solution (pH 9) is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing. It is also used in chemical analysis and in calibration of pH meters.