Decamethylcyclopentasiloxane, 97%
Decamethylcyclopentasiloxane, 97%
Decamethylcyclopentasiloxane, 97%
Decamethylcyclopentasiloxane, 97%
Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%
CAS: 541-02-6 | C10H30O5Si5 | 370.77 g/mol
Have Questions?
Catalog NumberQuantity
ALF042412.A11 kg
Catalog number ALF042412.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialDecamethylcyclopentasiloxane
CAS541-02-6
Health Hazard 1H227
Health Hazard 3P210-P280-P370+P378q-P501c
Melting Point-44°C
View more
Decamethylcyclopentasiloxane is employed in cosmetics like deodorants, sunblocks, hair sprays and personal care products. Furthermore, it is used in silicone based personal lubricants. In addition, it is a cyclic volatile methylsiloxane used in hair conditioners, thereby brushing the hair without breakage.