Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)
Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)
Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)
Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)
Thermo Scientific Chemicals
Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)
Coupling/cross-linking agent for precious metals | CAS: 40372-72-3 | C18H42O6S4Si2 | 538.94 g/mol
Have Questions?
Catalog NumberQuantity
ALF042410.22100 g
Catalog number ALF042410.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBis[3- (triethoxysilyl)propyl]tetrasulfide
CAS40372-72-3
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Bis[3-(triethoxysilyl)propyl]tetrasulfide is used in radial tires, rubber tire treads, carcass, tire walls, solid rubber tires and other rubber products. It acts as a chemical intermediate and as a cross linking agent. Furthermore, it acts as a coupling agent used to chemically modify silica surface. In addition, it serves as a curing agent for good heat aging.