Poly(2-ethyl-2-oxazoline), M.W. 500,000
Poly(2-ethyl-2-oxazoline), M.W. 500,000
Poly(2-ethyl-2-oxazoline), M.W. 500,000
Poly(2-ethyl-2-oxazoline), M.W. 500,000
Thermo Scientific Chemicals
Poly(2-ethyl-2-oxazoline), M.W. 500,000
CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF042398.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly (2-ethyl-2-oxazoline)
Name NoteM.W. 500,000
CAS25805-17-8
Physical FormOff-white solid
Recommended StorageAmbient temperatures
View more
Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.