Platinum foil, 0.127mm (0.005in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.127mm (0.005in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.127mm (0.005in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.127mm (0.005in) thick, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals
Platinum foil, 0.127mm (0.005in) thick, Premion™, 99.99% (metals basis)
Form: Approx.1.7g/25 x 25mm | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF042370.FI
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 x 50 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
CAS7440-06-4
AppearanceSilvery white foil