Silver foil, 0.1mm (0.004in) thick, annealed, Premion™, 99.998% (metals basis)
Silver foil, 0.1mm (0.004in) thick, annealed, Premion™, 99.998% (metals basis)
Silver foil, 0.1mm (0.004in) thick, annealed, Premion™, 99.998% (metals basis)
Silver foil, 0.1mm (0.004in) thick, annealed, Premion™, 99.998% (metals basis)
Thermo Scientific Chemicals
Silver foil, 0.1mm (0.004in) thick, annealed, Premion™, 99.998% (metals basis)
Form: approx. 2.6g/50x50mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
Catalog number ALF042317.GH
Price (MYR)
1,107.00
EA
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
1,107.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name NoteAnnealed
CAS7440-22-4
AppearanceSilver-colored foil