Gold plating solution, electroless, metal content≈3.7g/L
Gold plating solution, electroless, metal content≈3.7g/L
Thermo Scientific Chemicals
Gold plating solution, electroless, metal content≈3.7g/L
For coating nickel or copper
Have Questions?
Catalog NumberQuantity
ALF042307.AK250 mL
Catalog number ALF042307.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGold plating solution
ColorColorless to Yellow
Name NoteElectroless, metal content ≈3.7g/L
Health Hazard 1H311-H314-H332-H335
Health Hazard 2GHS H Statement
H301-H311-H331-H314
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
View more
For coating nickel or copper