Tri(1-naphthyl)phosphine, 97%
Tri(1-naphthyl)phosphine, 97%
Tri(1-naphthyl)phosphine, 97%
Tri(1-naphthyl)phosphine, 97%
Thermo Scientific Chemicals
Tri(1-naphthyl)phosphine, 97%
CAS: 3411-48-1 | C30H21P | 412.47 g/mol
Have Questions?
Catalog NumberQuantity
ALF042304.0910 g
Catalog number ALF042304.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTri (1-naphthyl)phosphine
CAS3411-48-1
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c