Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Thermo Scientific Chemicals
Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
ALF042286.SA2 Ea.
Catalog number ALF042286.SA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 Ea.
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl sulfoxide-d6
Density1.18 g/mL
Flash Point88°C (190°F)
OdorCharacteristic
PackagingPrescored Ampule
View more
DMSO is a common polar solvent used in NMR, PCR, as a cryoprotectant, and as a universal solvent for many compounds. Deuterated DMSO is a common solvent used in NMR spectroscopy.