Amberlite™ IRA-67, ion exchange resin
Amberlite™ IRA-67, ion exchange resin
Thermo Scientific Chemicals
Amberlite™ IRA-67, ion exchange resin
Acrylic-DVB; Weakly basic gel type resin | CAS: 80747-90-6 | Styrene-DVB
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ALF042253.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite IRA-200C(Na)
Physical FormBeads
Name NoteIon Exchange Resin
CAS80747-90-6
Recommended StorageAmbient temperatures
View more
Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.