Polyethylene oxide, M.W. 100,000
Polyethylene oxide, M.W. 100,000
Polyethylene oxide, M.W. 100,000
Polyethylene oxide, M.W. 100,000
Polyethylene oxide, M.W. 100,000
Thermo Scientific Chemicals
Polyethylene oxide, M.W. 100,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF042236.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene oxide
Name NoteM.W. 100,000
pH5 to 7
CAS25322-68-3
Physical FormPowder
View more
Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.