Silica, standard solution, Specpure™ SiO{2} 1000μg/mL
Silica, standard solution, Specpure™ SiO{2} 1000μg/mL
Thermo Scientific Chemicals
Silica, standard solution, Specpure™ SiO{2} 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF042225.AE100 mL
Catalog number ALF042225.AE
Price (MYR)
602.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
602.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilica standard solution
Name NoteSiO2 1000μg/mL
Recommended StorageAmbient temperatures
TSCAYes
Physical FormLiquid