Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Form: approx. 2.2g/50x50mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF042189.FI50 x 50 mm
Catalog number ALF042189.FI
Price (MYR)
624.00
EA
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
624.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
AppearanceCopper colored foil
Density8.94g/cm3