Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity
Widely used for proton exchange membrane (PEM) fuel cells and water electrolyzers. It is popular for separators in electrochemical cells requiring high ionic conductivity such as fuel cells and water electrolyzers. | CAS: 31175-20-9 | C9HF17O5S | 544.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
15 x 15 cm
30 x 30 cm
Catalog number ALF042180.VA
Price (MYR)
-
Quantity:
15 x 15 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialNafion™ N-117 membrane
CAS31175-20-9
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
Nafion™ N-117 membrane is widely used for proton exchange membrane (PEM) fuel cells and water electrolyzers. It is popular for separators in electrochemical cells requiring high ionic conductivity such as fuel cells and water electrolyzers. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.