Petroleum ether 35/60, ACS
Petroleum ether 35/60, ACS
Thermo Scientific Chemicals
Petroleum ether 35/60, ACS
Maximum level of impurities: Color (APHA): 10, Boiling range 35-60°C, Evaporation residue: 0.001%, Acidity P.T. | CAS: 8032-32-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF042085.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPetroleum ether 35/60
CAS8032-32-4
ColorColorless
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
View more
Petroleum ether is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of ficus thoningii.