Dodecanoic acid, typically 99.5%
Dodecanoic acid, typically 99.5%
Dodecanoic acid, typically 99.5%
Dodecanoic acid, typically 99.5%
Thermo Scientific Chemicals
Dodecanoic acid, typically 99.5%
CAS: 143-07-7 | C12H24O2 | 200.322 g/mol
Have Questions?
Catalog NumberQuantity
ALF042038.A11 kg
Catalog number ALF042038.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialDodecanoic acid
CAS143-07-7
Health Hazard 1H318
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P280-P305+P351+P338-P310-P501c
View more
Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.