Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Thermo Scientific Chemicals
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001% | CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF041986.22
Price (MYR)
1,598.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,598.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydrogen iodate
Name NotePrimary Standard
CAS13455-24-8
Health Hazard 1H272-H314-H335
Health Hazard 2Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
View more
Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.