Iodine, resublimed crystals, ACS, 99.8%
Iodine, resublimed crystals, ACS, 99.8%
Iodine, resublimed crystals, ACS, 99.8%
Iodine, resublimed crystals, ACS, 99.8%
Thermo Scientific Chemicals
Iodine, resublimed crystals, ACS, 99.8%
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF041955.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIodine
Melting Point183°C to 186° (sublimation)
CAS7553-56-2
ColorGray-Purple
Health Hazard 1H302+H312+H332-H315-H319-H335-H372