Molybdenum(IV) sulfide, 98%
Molybdenum(IV) sulfide, 98%
Molybdenum(IV) sulfide, 98%
Molybdenum(IV) sulfide, 98%
Thermo Scientific Chemicals
Molybdenum(IV) sulfide, 98%
CAS: 1317-33-5 | MoS2 | 160.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF041827.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMolybdenum(IV) sulfide
Melting Point1185°C
CAS1317-33-5
ColorBlack
Health Hazard 1H332
View more
Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.