Boron oxide, 99.999% (metals basis)
Boron oxide, 99.999% (metals basis)
Boron oxide, 99.999% (metals basis)
Boron oxide, 99.999% (metals basis)
Thermo Scientific Chemicals
Boron oxide, 99.999% (metals basis)
CAS: 1303-86-2 | B2O3 | 69.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF041786.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Boiling Point1860°C
Chemical Name or MaterialBoron oxide
Melting Point∽450°C
CAS1303-86-2
ColorWhite
View more
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.