Phthalic acid, ACS, 99.5+%
Phthalic acid, ACS, 99.5+%
Phthalic acid, ACS, 99.5+%
Phthalic acid, ACS, 99.5+%
Thermo Scientific Chemicals
Phthalic acid, ACS, 99.5+%
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Catalog NumberQuantity
ALF041770.36500 g
Catalog number ALF041770.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhthalic acid
CAS88-99-3
Melting Point205°C (decomposition)
Health Hazard 1H315-H318-H335
ColorWhite
View more
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.