Chloroacetic acid, ACS, 99%
Chloroacetic acid, ACS, 99%
Chloroacetic acid, ACS, 99%
Chloroacetic acid, ACS, 99%
Thermo Scientific Chemicals
Chloroacetic acid, ACS, 99%
CAS: 79-11-8 | C2H3ClO2 | 94.494 g/mol
Have Questions?
Catalog NumberQuantity
ALF041724.36500 g
Catalog number ALF041724.36
Price (MYR)
491.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
491.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroacetic Acid
CAS79-11-8
Health Hazard 1H301+H311-H314-H330-H335
Health Hazard 2GHS H Statement
H301-H311-H331-H314
Health Hazard 3P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
Chloroacetic acid is used in the production of pharmaceuticals, dyes, detergents, synthetic caffeine, glycine and thioglycolic acid, which acts as a stabilizer in PVC. It acts as a precursor to glyphosate and adrenaline. It also serves as a preservative and bacteriostat. Further, it is used as surface active agents and photosensitive chemicals. It is also used in the O-alkylation of salicylaldehyde and an active component in cosmetics. In addition, it is involved in the preparation of thickening agent carboxymethyl cellulose and carboxymethyl starch.