Zinc sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Zinc sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Zinc sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Zinc sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals
Zinc sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
ALF041655.KS1 Ea.
Catalog number ALF041655.KS
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc sputtering target
Name Note50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Quantity1 Ea.
CAS7440-66-6
OdorOdorless
View more
Magnetron sputtering source