Di-n-octyl phthalate, 98%
Di-n-octyl phthalate, 98%
Di-n-octyl phthalate, 98%
Di-n-octyl phthalate, 98%
Thermo Scientific Chemicals
Di-n-octyl phthalate, 98%
Plasticizer | CAS: 117-84-0 | C24H38O4 | 390.564 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Catalog number ALF041522.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-octyl phthalate
CAS117-84-0
Melting Point-25°C
ColorColorless to Yellow
Recommended StorageAmbient temperatures
View more
Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.