Copper Thinfoil, 0.015mm (0.0006in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Copper Thinfoil, 0.015mm (0.0006in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Copper Thinfoil, 0.015mm (0.0006in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Copper Thinfoil, 0.015mm (0.0006in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Copper Thinfoil, 0.015mm (0.0006in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF041504.FF25 x 25 mm
Catalog number ALF041504.FF
Price (MYR)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper Thinfoil
Name Note0.015mm (0.0006 in.) thick, Not certified pinhole free
CAS7440-50-8
Recommended StorageAmbient temperatures
EINECS Number231-159-6