Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis)
Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis)
Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis)
Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis)
Thermo Scientific Chemicals
Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis)
Form: Approx. 0.51g/m; Temper: hard | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF041455.G925 m
Catalog number ALF041455.G9
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 m
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name NoteHard
CAS7440-22-4
AppearanceSilver-colored wire