Xylenol Orange tetrasodium salt, ACS
Xylenol Orange tetrasodium salt, ACS
Xylenol Orange tetrasodium salt, ACS
Xylenol Orange tetrasodium salt, ACS
Thermo Scientific Chemicals
Xylenol Orange tetrasodium salt, ACS
CAS: 3618-43-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF041379.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialXylenol Orange tetrasodium salt
Melting Point210°C (decomposition)
CAS3618-43-7
Recommended StorageAmbient temperatures
EINECS Number222-805-8
View more
Xylenol Orange tetrasodium salt is used as a photometric reagent and a metal indicator. It is a versatile indicator for EDTA titrations in acidic range. It is a common organic reagent used as an indicator for metal titrations.