Nickel wire, 1.0mm (0.04in) dia, hard, 99.5% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, hard, 99.5% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, hard, 99.5% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, hard, 99.5% (metals basis)
Thermo Scientific Chemicals
Nickel wire, 1.0mm (0.04in) dia, hard, 99.5% (metals basis)
CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF041361.H250 m
Catalog number ALF041361.H2
Price (MYR)
1,622.00
EA
Add to cart
Quantity:
50 m
Request bulk or custom format
Price (MYR)
1,622.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel wire
Name Note1.0mm (0.04 in.) dia., Hard, Temper: as drawn
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.