Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight
Thermo Scientific Chemicals
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF041238.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinyl alcohol
Name NoteLow molecular weight, Hydrolyzed
CAS9002-89-5
Recommended StorageAmbient temperatures
TSCAYes
View more
Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.