2-Propanol, GC Grade, 99.7+%
2-Propanol, GC Grade, 99.7+%
2-Propanol, GC Grade, 99.7+%
2-Propanol, GC Grade, 99.7+%
Thermo Scientific Chemicals
2-Propanol, GC Grade, 99.7+%
Suitable For Environmental, Pesticide, and GC Analysis | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog NumberQuantity
ALF040983.K21 L
Catalog number ALF040983.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material2-Propanol
Melting Point°C to 90°C
Name NoteGC Grade
CAS67-63-0
ColorColorless
View more
Suitable for environmental and GC analysis. Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, anti freezes, soaps and window cleaners.