Ethyl acetate, Environmental Grade, 99.5+%
Ethyl acetate, Environmental Grade, 99.5+%
Ethyl acetate, Environmental Grade, 99.5+%
Ethyl acetate, Environmental Grade, 99.5+%
Thermo Scientific Chemicals
Ethyl acetate, Environmental Grade, 99.5+%
Pesticide residue, environmental, and GC analysis | CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog NumberQuantity
ALF040977.K21 L
Catalog number ALF040977.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-84°C
CAS141-78-6
ColorColorless
Health Hazard 1H225-H319-H336
View more
Ethyl acetate is used primarily as a solvent and diluent. It is commonly used to clean the circuit boards, in some nail varnish removers and also in residue, environmental and GC analysis. It is used chiefly as a scent in the manufacture of perfumes. It is also used in flavorings, glues, in decaffeinating tea and coffee, and in cigarettes. The other minor applications of ethyl acetate include its applications in the textiles, dyestuffs, explosives, photographic films and plates, artificial leather.