Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals
Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF040760.GH100 x 100 mm
Catalog number ALF040760.GH
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 x 100 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum foil
Name Note0.1mm (0.004 in.) thick, Temper: as rolled
CAS7429-90-5
Recommended StorageAmbient temperatures
EINECS Number231-072-3