Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 98%
Reducing agent, specific for disulfides | CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol
Have Questions?
Catalog NumberQuantity
ALF040587.042 g
Catalog number ALF040587.04
Price (MYR)
794.00
EA
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (MYR)
794.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris(2-carboxyethyl)phosphine hydrochloride
CAS51805-45-9
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Reducing agent, specific for disulfidesTris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides, and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.