Holmium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Holmium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Holmium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Holmium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Thermo Scientific Chemicals
Holmium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
CAS: 14017-54-0 | Cl3HoO12 | 463.268 g/mol
Have Questions?
Catalog NumberQuantity
ALF040575.NC10 g
Catalog number ALF040575.NC
Price (MYR)
1,660.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
1,660.00
EA
Add to cart
Specifications
Chemical Name or MaterialHolmium(III) perchlorate
CAS14017-54-0
Health Hazard 1H271-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Holmium(III) perchlorate is used to verify the wavelength scale of UV/visible spectrophotometers.