Bismuth Lead Tin ingot (Rose's metal)
Bismuth Lead Tin ingot (Rose's metal)
Bismuth Lead Tin ingot (Rose's metal)
Bismuth Lead Tin ingot (Rose's metal)
Thermo Scientific Chemicals
Bismuth Lead Tin ingot (Rose's metal)
CAS: 56001-37-7 | C10H14N5Na2O14P3 | 567.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF040516.36500 g
Catalog number ALF040516.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth Lead Tin ingot (Rose's metal)
CAS56001-37-7
Melting Point95°C to 96°C
Health Hazard 1H302+H332-H360Df-H373
Health Hazard 2GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
View more
Rose's metal was used as the droplet fluid, because it has a low melting point (e460 K). Rose's metal is used as a sample in computer-controlled diffractometer.