Ammonium sulfide, 40-44% w/w aq. soln.
Ammonium sulfide, 40-44% w/w aq. soln.
Ammonium sulfide, 40-44% w/w aq. soln.
Ammonium sulfide, 40-44% w/w aq. soln.
Thermo Scientific Chemicals
Ammonium sulfide, 40-44% w/w aq. soln.
CAS: 12135-76-1 | H8N2S | 68.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF040509.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium sulfide
CAS12135-76-1
ColorYellow to Orange
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth.It is an unstable salt and acts as a selective reducing agent.